Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamidine acetate, 99%

Catalog No. p-7081261
Click to view available options
Quantity:
100 g
500 g

CAS: 3473-63-0 | C3H8N2O2 | 104.109 g/mol

Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

Solubility
Soluble in water and hot alcohol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3473-63-0
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD00012866
InChI Key XPOLVIIHTDKJRY-UHFFFAOYSA-N
Synonym formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
PubChem CID 160693
IUPAC Name acetic acid;methanimidamide
SMILES CC(=O)O.C(=N)N

Specifications

Melting Point ∼158°C (decomposition)
Density 1.124
pH 8
Quantity 100 g
Beilstein 3563488
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Formamidine acetate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 222-442-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.