Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamidinesulfinic acid, 99%

Catalog No. p-136826
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1758-73-2 | CH4N2O2S | 108.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-73-2
Molecular Formula CH4N2O2S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00002397
InChI Key FYOWZTWVYZOZSI-UHFFFAOYSA-N
Synonym thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
PubChem CID 61274
IUPAC Name amino(imino)methanesulfinic acid
SMILES C(=N)(N)S(=O)O

Specifications

Melting Point 126.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NC(=NH)SO2H
Quantity 10 g
Beilstein 03, I, 36
Fieser 04,506; 05,668; 06,586
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents
Formula Weight 108.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Formamidinesulfinic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Self-heating in large quantities; may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
May cause damage t
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep cool.
Protect from sunlight.
Avoid breathing dust/fume/ga
EINECSNumber 217-157-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.