Learn More
Mopec™ Formaway Formalin Neutralizing Powder
SDP
Catalog No. p-200008025 Shop All Mopec Products
Click to view available options
Weight (English):
14 lb.
38 lb.
Formaway Formalin Neutralizing Powder safely and easily neutralizes spent formalin waste to a non-hazardous and non-toxic solution in only 20 minutes. Contents are emptied directly into 1 gallon of spent 10% Formalin or 4% Glutaldehyde, reducing risk exposure to lab personnel.
This unique formula also eliminates drain-clogging polymers.
  • Collect waste in a designated collection container
  • Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
  • Add the entire contents of one Formaway bottle to each gallon of aldehyde waste
  • Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of Formaway Formalin Neutralizer Powder
  • Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
  • If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for Formaway Formaldehyde Residual Screening Test Kit)
  • Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for re-use
Specifications
Material Pre-treated Formalin Neutralizer
Type Formalin Neutralizer
Packaging Quantity 12 Bottles/Cs.
Weight (English) 14 lb.
Weight (Metric) 6.35 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.