Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Formic Acid, 88%
SCI_ED

Catalog No. s25331 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-18-6, 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Color Colorless
Assay Percent Range 88% w/v
Linear Formula HCOOH
Formula Weight 46.03
Grade Reagent
Density 1.22g/mL
Quantity 100 mL
Packaging Reagent Grade
Chemical Name or Material Formic Acid, 88%
ShelfLife 1 year
DOTInformation DOT Class 8 (3), II, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.