Learn More
Formic acid, ACS, 88+%
Description
It is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 7°C to 9°C |
Boiling Point | 100°C to 101°C |
Linear Formula | HCOOH |
UN Number | 1779 |
Beilstein | 1209246 |
Merck Index | 14,4241 |
Solubility Information | Miscible in water,ether,acetone,ethyl acetate,glycerol,methanol and ethanol. Partially miscible in benzene,toluene and xylene. |
Formula Weight | 46.03 |
Percent Purity | ≥88% |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H226-H302-H314-H331 |
Hazard Statement | GHS H Statement H331-H314-H318-H226-H302 |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P363-P372-P374-P380-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID |
EINECSNumber | 200-579-1 |
RTECSNumber | LQ4900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.