Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, FCC, 85%, Spectrum™ Chemical
SDP

Catalog No. 18602847 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-847 6 x 500 mL Amber Glass Bottle
18-602-846 500 mL Amber Glass Bottle
18-602-850 2.5 L Glass Bottle
18-602-851 4 x 2.5 L Glass Bottle
18-602-845 20 L Poly Drum
18-602-852 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-847 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1135500MLGLCS6
Add to Cart
Add to Cart

F1135, 64-18-6, CH2O2

Spectrum™ Chemical Formic Acid, FCC is the simplest carboxylic acid and one of its biggest uses is as a preservative and antibacterial agent in livestock feed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Max % 100%
UN Number UN1779
Percent Purity 85%
Grade FCC
Quantity 6 x 500 mL
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.