Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, FCC, 85%, Spectrum™ Chemical
SDP

Catalog No. p-7157440
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum

F1135, 64-18-6, CH2O2

Spectrum™ Chemical Formic Acid, FCC is the simplest carboxylic acid and one of its biggest uses is as a preservative and antibacterial agent in livestock feed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Max % 100%
UN Number UN1779
Percent Purity 85%
Grade FCC
Quantity 500 mL
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.