Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Solstice

Catalog No. 6000623 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
70 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-23 2.5 L
60-006-22 1 L
60-006-25 70 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-006-23 Supplier Solstice Supplier No. 270012.5L

Puriss., meets analytical specifications of DAC, FCC, 98.0-100%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Linear Formula HCOOH
UN Number 1779
Beilstein 1209246
Formula Weight 46.03g/mol
Percent Purity 98 to 100%
Grade puriss.
Density 1.22g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.57
Flash Point 48°C (118.4°F)
Ignition Point 1004°F
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8mmHg (20°C)
Chemical Name or Material Formic Acid, Meets analytical specifications of DAC, FCC
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
Precautionary Statement H226-H302-H314-H331
EINECSNumber 200-579-1
RTECSNumber LQ4900000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.