Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid, 88%, ACS reagent

Catalog No. p-4509568
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 8.0°C
Boiling Point 101.0°C
Assay Percent Range 88%
Linear Formula HCO2H
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine
Formula Weight 46.02
Percent Purity 88%
Grade ACS Reagent
Density 1.2200g/mL
Quantity 500 mL
Flash Point 60°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Liquid
Refractive Index 1.3711 to 1.3731
Residue after Evaporation 0.002% max.
Specific Gravity 1.22
Chemical Name or Material Formic acid
Show More Show Less
Hazard Category Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.