Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Formoterol hemifumarate
GREENER_CHOICE

Catalog No. 144810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-481-0 10 mg
14-485-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-481-0 Supplier Tocris Bioscience™ Supplier No. 1448/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective β2 agonist

Formoterol hemifumarate is a potent, selective and long-acting β2-adrenoceptor agonist. Displays 330-fold selectivity for β2 over β1 receptors (pKd values are 8.12 and 5.58 respectively). Potently relaxes guinea pig trachea (pD2 = 9.29), and is longer-acting and 100-fold more potent than Salbutamol (Cat. No. 0634).

Chemical Identifiers

CAS 43229-80-7
Molecular Formula C42H52N4O12

Specifications

Quantity 10 mg
Formula Weight 402.45
Purity >99%
Molecular Weight (g/mol) 804.894
Chemical Name or Material Formoterol hemifumarate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.