Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Formyl-Histone H1.2 (Lys74) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144348
Description
Formyl-Histone H1.2 (Lys74) Polyclonal Antibody for ChIP, ELISA
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
Formyl-Histone H1.2 (Lys74) | |
Polyclonal | |
Unconjugated | |
H1-2 | |
0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3006 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, ELISA | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P16403 | |
H1-2, HIST1H1C | |
Peptide sequence around site of Formyl-Lys (74) derived from Human Histone H1.2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction