Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Formyl-Histone H2B (Lys116) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112476 Supplier Invitrogen™ Supplier No. PA5112476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Formyl-Histone H2B (Lys116) Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Formyl-Histone H2B (Lys116)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene H2BW2
Gene Accession No. A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880
Gene Alias 2610022J01Rik; Acetyl-Histone H2; AV127319; bA317E16.3; CG17949; CG17949-PA; CG33868; CG33868-PA; CG33870; CG33870-PA; CG33872; CG33872-PA; CG33874; CG33874-PA; CG33876; CG33876-PA; CG33878; CG33878-PA; CG33880; CG33880-PA; CG33882; CG33882-PA; CG33884; CG33884-PA; CG33886; CG33886-PA; CG33888; CG33888-PA; CG33890; CG33890-PA; CG33892; CG33892-PA; CG33894; CG33894-PA; CG33896; CG33896-PA; CG33898; CG33898-PA; CG33900; CG33900-PA; CG33902; CG33902-PA; CG33904; CG33904-PA; CG33906; CG33906-PA; CG33908; CG33908-PA; CG33910; CG33910-PA; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; Dmel\CG17949; Dmel\CG33868; Dmel\CG33870; Dmel\CG33872; Dmel\CG33874; Dmel\CG33876; Dmel\CG33878; Dmel\CG33880; Dmel\CG33882; Dmel\CG33884; Dmel\CG33886; Dmel\CG33888; Dmel\CG33890; Dmel\CG33892; Dmel\CG33894; Dmel\CG33896; Dmel\CG33898; Dmel\CG33900; Dmel\CG33902; Dmel\CG33904; Dmel\CG33906; Dmel\CG33908; Dmel\CG33910; Dmel_CG17949; Dmel_CG33868; Dmel_CG33870; Dmel_CG33872; Dmel_CG33874; Dmel_CG33876; Dmel_CG33878; Dmel_CG33880; Dmel_CG33882; Dmel_CG33884; Dmel_CG33886; Dmel_CG33888; Dmel_CG33890; Dmel_CG33892; Dmel_CG33894; Dmel_CG33896; Dmel_CG33898; Dmel_CG33900; Dmel_CG33902; Dmel_CG33904; Dmel_CG33906; Dmel_CG33908; Dmel_CG33910; fj05g08; GL105; H2B; H2B 291A; H2B 291B; H2b 613; H2B clustered histone 1; H2B clustered histone 15; H2B clustered histone 17; H2B clustered histone 19; H2B clustered histone 2, pseudogene; H2B clustered histone 21; H2B clustered histone 5; H2B clustered histone 8; H2B GL105 antibody; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B I; H2B II; H2B III; H2B IV; H2B K; H2B type 12; H2B VI; H2B VII; H2B.1; H2B.1 antibody; H2B.1A; H2B.1B; H2B.2; H2B.U histone 1; H2B.W histone 1; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/I; H2B/j; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/r; H2B/S; H2B/t; H2B1; H2B1.1; H2B1.2; h2B-143; H2B2; H2b-221; H2b-291b; H2b-613; H2Bb; H2BC1; H2BC10; H2BC15; H2BC17; H2BC19; H2BC21; H2BC2P; H2BC4; H2BC5; H2BC6; H2BC7; H2BC8; H2b-f; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFO; H2BFQ; h2bfr; H2BFS; H2BFT; H2BFU; H2BFWT; H2BGL105; H2B-I; H2B-II; H2B-III; H2B-IV; H2BJ; H2b-j; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2b-l; H2b-n; H2BQ; H2BS14; H2BS14p; H2BU1; H2B-VI; H2B-VII; H2BW1; HIRA-interacting protein 1; HIRA-interacting protein 2; HIRIP1; HIRIP2; His2B; His2B:CG17949; His2B:CG17949-PA; His2B:CG33868; His2B:CG33868-PA; His2B:CG33870; His2B:CG33870-PA; His2B:CG33872; His2B:CG33872-PA; His2B:CG33874; His2B:CG33874-PA; His2B:CG33876; His2B:CG33876-PA; His2B:CG33878; His2B:CG33878-PA; His2B:CG33880; His2B:CG33880-PA; His2B:CG33882; His2B:CG33882-PA; His2B:CG33884; His2B:CG33884-PA; His2B:CG33886; His2B:CG33886-PA; His2B:CG33888; His2B:CG33888-PA; His2B:CG33890; His2B:CG33890-PA; His2B:CG33892; His2B:CG33892-PA; His2B:CG33894; His2B:CG33894-PA; His2B:CG33896; His2B:CG33896-PA; His2B:CG33898; His2B:CG33898-PA; His2B:CG33900; His2B:CG33900-PA; His2B:CG33902; His2B:CG33902-PA; His2B:CG33904; His2B:CG33904-PA; His2B:CG33906; His2B:CG33906-PA; His2B:CG33908; His2B:CG33908-PA; His2B:CG33910; His2B:CG33910-PA; HIST1H2B7; HIST1H2B7L3; hist1h2ba; HIST1H2BB; HIST1H2BB antibody; Hist1h2bc; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; Hist1h2bh; HIST1H2BI; Hist1h2bj; hist1h2bj.L; hist1h2bj.S; HIST1H2BK; HIST1H2BL
Gene Symbols H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, LOC102724334
Host Species Rabbit
Immunogen Peptide sequence around site of Formyl-Lys (116) derived from Human Histone H2B type 1-C/E/F/G/I (112-123aa).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102724334, 114483833, 128312, 158983, 255626, 286436, 3017, 3018, 440689, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.