Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Forsterite, Thermo Scientific™

Catalog No. AA4263914
Click to view available options
Quantity:
25 g

Catalog No. AA4263914

Supplier: Thermo Scientific Chemicals 4263914

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15118-03-3
Molecular Formula H4Mg2O4Si
Molecular Weight (g/mol) 144.72
MDL Number MFCD00283278
InChI Key ICUHFTWXYNJXLX-UHFFFAOYSA-N
IUPAC Name silicic acid dimagnesium
SMILES [Mg].[Mg].O[Si](O)(O)O

Specifications

Color Green
Physical Form Grains
Quantity 25 g
Solubility Information Soluble in hot HCl. Insoluble in water.
Formula Weight 140.71
Odor Odorless
Refractive Index 1.63
Appearance Light green chunks
Chemical Name or Material Forsterite
Hazard Category H332: Harmful if inhaled.
EINECSNumber 239-169-2
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.