Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOS, Rabbit anti-Human, Clone: A16-L, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15927
Specifications
FOS | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
AP-1/C-FOS | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
A16-L | |
ELISA (1:20000-1:50000) Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
v-fos FBJ murine osteosarcoma viral oncogene homolog | |
FOS | |
A synthetic peptide corresponding to amino acids 358-372 at C-terminus of human FOS. | |
RUO | |
2353 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction