Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOS, Rabbit anti-Human, Clone: F14-R, Abnova™

Catalog No. 89998724 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-724 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-724 Supplier Abnova Corporation Supplier No. MAB15926
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen FOS
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F14-R
Conjugate Unconjugated
Dilution ELISA (1:20000-1:50000) Immunocytochemistry Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide).
Gene v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Alias AP-1/C-FOS
Gene Symbols FOS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 4-16 at N-terminus of human FOS.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2353
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.