Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FosB/G0S3 Antibody (83B1138), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056530AF532
Description
FosB/G0S3 Monoclonal specifically detects FosB/G0S3 in Human samples. It is validated for Western Blot.Specifications
FosB/G0S3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
FOSB | |
This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
83B1138 | |
Western Blot | |
activator protein 1, AP-1, DKFZp686C0818, FBJ murine osteosarcoma viral oncogene homolog B, G0S3G0/G1 switch regulatory protein 3, GOS3, GOSB, MGC42291, oncogene FOS-B, protein fosB | |
Mouse | |
Protein G purified | |
Cancer, Tumor Suppressors | |
2354.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction