Learn More
FOSB Mouse anti-Human, Clone: OTI5H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804194

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
FOSB | |
Monoclonal | |
Unconjugated | |
FOSB | |
AP-1, G0S3, GOS3, GOSB | |
Mouse | |
Affinity Chromatography | |
RUO | |
2354 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P53539 | |
FOSB | |
Human recombit protein fragment corresponding to amino acids 1-255 of human FOSB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.