Learn More
Invitrogen™ FosB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579280
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat thymus tissue, mouse thymus tissue, mouse ovary tissue, mouse spleen tissue, mouse kidney tissue, mouse RAW2467 whole cell, human HL-60 whole cell, human THP-1 whole cell. IHC: human renal cancer tissue, human placenta tissue, mouse intestine tissue, rat intestine tissue. Flow: SiHa cell.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
Specifications
FosB | |
Polyclonal | |
Unconjugated | |
Fosb | |
activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100360880, 14282, 2354 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
D3ZLB7, P13346, P53539 | |
Fosb | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Fos B (264-291aa LPFQTSQDAPPNLTASLFTHSEVQVLGD). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.