Learn More
Invitrogen™ FosB Recombinant Rabbit Monoclonal Antibody (6M9D5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558100

Description
Immunogen sequence: MFQAFPGDYD SGSRCSSSPS AESQYLSSVD SFGSPPTAAA SQECAGLGEM PGSFVPTVTA ITTSQDLQWL VQPTLISSMA QSQGQPLASQ PPVVDPYDMP.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
Specifications
FosB | |
Recombinant Monoclonal | |
2.16 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
D3ZLB7, P53539 | |
Fosb | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Fos B (NP_006723.2). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
6M9D5 | |
Unconjugated | |
Fosb | |
activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
Rabbit | |
Protein A | |
RUO | |
100360880, 2354 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.