Learn More
Foss™ Accessories for Soxtec™ Fat Extraction Systems: Aluminum Extraction Cups
Click to view available options
For Use With (Equipment):
Soxtec 2050 and 2055
Soxtec Unit HT 6, HT 12, 2043
Description
†For environmental applications, such as PCB extraction.
Specifications
Specifications
| For Use With (Equipment) | Soxtec 2050 and 2055 |
| Product Type | Extraction Cup |
| Material | Aluminum |
| For Use With (Application) | Fat extraction |