Learn More
Foss™ Condenser Seals for Soxtec™ Extraction Systems
Supplier:  Foss™ 10005714
 View more versions of this product
Catalog No. TC10005714
Description
Description
Specifications
Specifications
Condenser Seal
Soxtec 2045 and 1045
PTFE Resin
For use with acetone, carbon tetrachloride, chloroform, chloroform-methanol (2:1), cyclohexane, dichloromethane, ethanol, ethyl acetate, heptane, hexane, methanol, petrole solvents
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.