Learn More
Foss™ Condenser Seals for Soxtec™ Extraction Systems
Catalog No. p-4704625
Click to view available options
For Use With (Equipment):
Soxtec 2045 and 1045
Material:
PTFE Resin
Specifications
Product Type Condenser Seal
For Use With (Equipment) Soxtec 2045 and 1045
Material PTFE Resin
For Use With (Application) For use with acetone, carbon tetrachloride, chloroform, chloroform-methanol (2:1), cyclohexane, dichloromethane, ethanol, ethyl acetate, heptane, hexane, methanol, petrole solvents
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.