Learn More
Foss™ Condenser Seals for Soxtec™ Extraction Systems
Supplier: Foss™ 10007526
Specifications
Condenser Seal | |
Soxtec 2050 and 2055 | |
Butyl | |
For use with acetone, acetonitrile, ethanol, ethyl acetate and methanol solvents |