Learn More
Foss™ Dummy Tubes
Catalog No. TC100235
Change view
Click to view available options
Quantity:
5/Pk.
Catalog No. Quantity
TC100235 5/Pk.
1 options
Catalog No. TC100235 Supplier Foss™ Supplier No. 100235
Specifications
Product Type Dummy Tube
For Use With (Equipment) Exhaust manifold when less than 8 or 20 samples are being digested
Quantity 5/Pk.