Learn More
Fotodyne™ FOTO/Analyst™ Investigator/FX Workstations with Manual Zoom, Side Lights
Catalog No. 1630F2DL
Change view
Click to view available options
Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
Dual-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Dual-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
16-30F-2DL Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx camera
1630F2DLNP Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-30F-2UV Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
1630F2UVNP Dual-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-70F-2DL Dual-wavelength UV dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx camera
1670F2DLNP Dual-wavelength UV dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
16-70F-2UV Dual-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
1670F2UVNP Dual-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 16-30F-2DL Supplier Fotodyne™ Supplier No. 71630F2DL
Includes
8-48mm (6X) manual zoom lens, manual filter cassette, choice of CCD camera, choice of transilluminator, FOTO/FX darkroom with dual white-light and UV epi-illumination, image acquisition software, PCI frame grabber (with 0.4 megapixel camera) or firewire (with 1.4 and 2.0 megapixel cameras) card, ethidium bromide filter, Coomassie™ blue filter (dual-light workstations only), PC, optional thermal printer, all necessary cables and connectors
Large-format transilluminators provide the flexibility to capture images of many different sample types, including gels, microplates, fluorescent blots, colorimetric blots, TLC plates, and bacterial plates
  • System is designed to be easily upgraded to include a cooled CCD camera for low-light samples
  • Files can be archived in several formats to preserve data integrity for downstream analysis
  • Workstation can be networked to make files more accessible
  • Manual zoom lens allows adjustments to be made by rotating the on-lens aperature, zoom, and focus rings
FOTO/FX Benchtop Darkroom
  • Light-tight enclosure houses a CCD camera with zoom lens and filters
  • Flat black interior minimizes stray light
  • Gliding transilluminator platform and side-opening door with interlock provide convenient, safe operation of large-format transilluminators
  • Two interior visible epi-light panels allow easy focusing and even illumination of opaque samples
  • Dual UV epi-lights can be used to optimize sensitivity of samples stained with fluorescent dyes
  • Camera mounting system allows for easy upgrading to other cameras
  • Front-loading filter holder allows simple exchange of filters
FOTO/Flex Transillumators
  • Choose single-wavelength UV; single-wavelength UV, dual-light (UV plus white light); dual-wavelength UV; or dual-wavelength UV, dual-light models
  • UV transilluminators have 25 × 28cm UV glass; four 8 watt, 312nm (peak emission) bulbs; UV transparent protective cover; and preparative and analytical intensity settings
  • White-light transilluminators have a proprietary electroluminescent (EL) visible light panel that offers 15 times more uniform light illumination than traditional bulb-style transilluminators and continuously adjustable intensity control
Image Acquisition Software
  • Integration to 8.3 min. in 1/30 sec. increments
  • Live preview
  • Brightness and contrast
  • Image manipulation: invert, zoom, mirror, rotate, annotate (date, time, text, circle, arrow)
  • Image storage in choice of TIF, JPG, BMP, and PNG formats
Additional configurations are available. Contact your Fisher Scientific Customer Service Representative for details.