Learn More
Fotodyne™ FOTO/Analyst™ Investigator/FX Workstations with Motorized Zoom, Side Lights, Single Wavelength
Catalog No. 1620DL
Change view
Click to view available options
Dual-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
Dual-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx
Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
16-20D-L Dual-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
16-20D-LNP Dual-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
16-20F-DL Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
16-20F-DLNP Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-20F-UV Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
16-20F-UVNP Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-20U-V UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
16-20U-VNP UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
71660DLNP Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera
16-60F-DL Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx
16-60F-DLNP Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
16-60F-UV Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
16-60F-UVNP Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
71660UVNP UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 16-20D-L Supplier Fotodyne™ Supplier No. 71620DL
Includes
11.5-69mm (6X) motorized zoom lens, motorized filter wheel, choice of CCD camera, choice of transilluminator, FOTO/FX darkroom with dual white-light and UV epi-illumination, image acquisition software, PCI frame grabber (with 0.4 megapixel camera) or firewire (with 1.4 and 2.0 megapixel cameras) card, Coomassie™ blue filter (dual-light workstations only), PC, optional thermal printer, all necessary cables and connectors
Large-format transilluminators provide the flexibility to capture images of many different sample types, including gels, microplates, fluorescent blots, colorimetric blots, TLC plates, and bacterial plates
  • Systems are designed to be easily upgraded to include a cooled CCD camera for low-light samples
  • Files can be archived in several formats to preserve data integrity for downstream analysis
  • Workstations can be networked to make files more accessible
  • Motorized zoom lens allows the camera lens and darkroom to be controlled through user-friendly software
  • All systems are PC-compatible
FOTO/FX Benchtop Darkroom
  • Light-tight enclosure houses a CCD camera with zoom lens and filters
  • Flat black interior minimizes stray light
  • Gliding transilluminator platform and side-opening door with interlock provide convenient, safe operation of large-format transilluminators
  • Two interior visible epi-light panels allow easy focusing and even illumination of opaque samples
  • Dual UV epi-lights can be used to optimize sensitivity of samples stained with fluorescent dyes
  • Camera mounting system allows for easy upgrading to other cameras
  • Motorized filter wheel permits filters to be selected through computer software or the darkroom touchpad
FOTO/Flex Transillumators
  • Choose single-wavelength UV; single-wavelength UV, dual-light (UV plus white light); dual-wavelength UV; or dual-wavelength UV, dual-light models
  • UV transilluminators have 25 × 28cm UV glass; four 8 watt, 312nm (peak emission) bulbs; UV transparent protective cover; and preparative and analytical intensity settings
  • White-light transilluminators have a proprietary electroluminescent (EL) visible light panel that offers 15 times more uniform light illumination than traditional bulb-style transilluminators and continuously adjustable intensity control
Image Acquisition Software
  • Integration to 8.3 min. in 1/30 sec. increments
  • Live preview
  • Brightness and contrast
  • Image manipulation: invert, zoom, mirror, rotate, annotate (date, time, text, circle, arrow)
  • Image storage in choice of TIF, JPG, BMP, and PNG formats