Learn More
Fotodyne™ FOTO/Analyst™ Investigator Workstations
Catalog No. F61710PDLNP
Change view
Click to view available options
Dual light workstation w/8-bit megapixel camera
UV workstation w/8-bit megapixel camera
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
F61710PDLNP Dual light workstation w/8-bit megapixel camera
F61710PUVNP UV workstation w/8-bit megapixel camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F61710PDLNP Supplier Fotodyne™ Supplier No. 61710PDLNP
Includes
  • FOTO/Eclipse™ benchtop darkroom
  • CCD camera
  • Transilluminator
  • Image acquisition software
  • Filters
High quality digital photographs of DNA, RNA, and protein gels
Versatile systems combine UV or dual-light transilluminator, benchtop darkroom, and CCD camera for seamless gel documentation.
  • Preview images and adjust camera settings while viewing real-time changes on a full-size computer monitor
  • Save images in a variety of file formats, including TIF, BMP, and JPG
  • Flexible integration offers wide range of adjustable exposure settings in 1/30 sec. increments
  • Nanogram-level sensitivity
  • FOTO/Eclipse benchtop darkroom features dual-white light epi-illumination
  • Gliding transilluminator drawer for easy gel positioning, cutting out bands, cleaning
  • Safety-interlocking door prevents accidental UV exposure
  • FOTO/Convertible Dual Light Transilluminator offers 21 × 26cm UV viewing surface covered by hinged electroluminescent white light panel
  • 8-bit, 0.4 megapixel camera is ideal for basic gel documentation, or choose an 8- or 12-bit 1.4 megapixel camera for more demanding applications
Order Info
Electrical Requirements: 120V 60Hz