Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Four-Station Table
SCI_ED

Catalog No. p-7121407 Shop All Diversified Spaces Products
Click to view available options
Product Type:
4-Station Table with Maple Top
4-Station Table with Plastic Laminate Top

Designed to provide structurally sound and completely functional work center

  • Constructed from solid maple and maple veneer
  • 2.063 x 11.43cm (0.8125 x 4.5 in.) solid maple aprons with 13-gauge steel corner braces
  • 5.715cm (2.25 in.) maple legs with adjustable floor glides and rubber leg boots
  • SEFA compliant and MAS Certified Green
  • Finished with durable chemical resistant non-emitting UV finish

Specifications

Product Type 4-Station Table with Maple Top
Dimensions (D x W x H) 121.92 x 121.92 x 76.2 cm (48 x 48 x 30 in.)
Thickness (English) 1.75 in.
Warranty Limited Lifetime
Color Maple
Thickness (Metric) 44.4 mm
Material Maple
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.