Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOX2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040588AF594
Description
FOX2 Polyclonal antibody specifically detects FOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FOX2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
dJ106I20.3, Fox-1 homolog B, fox-1 homologue, FOX2Fox-2, fxh, hexaribonucleotide binding protein 2, Hexaribonucleotide-binding protein 2, HRNBP2FOX-2, RBM9RTAHNRBP2, Repressor of tamoxifen transcriptional activity, RNA binding motif protein 9, RNA binding protein fox-1 homolog 2, RNA binding protein, fox-1 homolog (C. elegans) 2, RNA-binding motif protein 9, RNA-binding protein 9 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human RNA Binding Motif Protein 9 using the numbering given in entry NP_055124.1 (GeneID 23543). | |
0.1 mL | |
Cell Cycle and Replication | |
23543 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction