Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOX3/NeuN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983958
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3958 Supplier Bioss Supplier No. BS1613R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

Specifications

Antigen FOX3/NeuN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RBFOX3
Gene Accession No. A6NFN3
Gene Alias BOS_19554; D11Bwg0517e; Fox-1 homolog C; FOX3; Fox-3; hexaribonucleotide binding protein 3; Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3
Gene Symbols RBFOX3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NeuN.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146713
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.