Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXA1 Recombinant Rabbit Monoclonal Antibody (HL1974)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547244
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Specifications
FOXA1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P55317 | |
FOXA1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXA1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
HL1974 | |
Unconjugated | |
FOXA1 | |
Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2 | |
Rabbit | |
Protein A | |
RUO | |
3169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction