Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA1 Mouse anti-Human, Mouse, Clone: 3A8, Abnova™

Catalog No. 89035465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-465 Supplier Abnova Corporation Supplier No. MAB14060

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human FOXA1.

  • High specificity
  • High reactivity

Specifications

Antigen FOXA1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3A8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human FOXA1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P55317
Gene Alias HNF3A/MGC33105/TCF3A
Gene Symbols FOXA1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-473 of human FOXA1.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3169
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.