Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590342 Supplier Invitrogen™ Supplier No. PA590342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKRFKCEKQP GAGGGGGSGS GGSGAKGGPE SRKDPSGASN PSADSPLHRG VHGKTGQLEG APAPGPAASP QTLDHSGATA TGGASELKTP ASSTAPPISS GPGALASVPA SHPAHGLAPH ESQLHLKGDP HYSFNHPFSI NNLMSSSEQQ HKLDFKAYEQ ALQYSPYGST LPASLPLGSA SVTTRSPIEP SALEPAYYQG VYSRPVLNTS; Positive Samples: A-549, MCF7, LO2, HeLa, Mouse lung; Cellular Location: Nucleus.

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FOXA1
Gene Accession No. P23512, P35582, P55317
Gene Alias Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2
Gene Symbols FOXA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 263-472 of human FOXA1 (NP_0044872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15375, 25098, 3169
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.