Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA1 Rabbit anti-Human, Unconjugated, Clone: FOXA1/2230R, Abnova™

Catalog No. 89337857 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-857 Supplier Abnova Corporation Supplier No. RAB00432
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

FOXA1 Monoclonal antibody specifically detects FOXA1 in Human samples. It is validated for IHC-P.

Specifications

Antigen FOXA1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FOXA1/2230R
Conjugate Unconjugated
Description forkhead box A1
Gene FOXA1
Gene Alias HNF3A, MGC33105, TCF3A
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acid 372-472 of human FOXA1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3169
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.