Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA2/HNF 3beta Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984662
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4662 Supplier Bioss Supplier No. BS2358R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.

Specifications

Antigen FOXA2/HNF 3beta
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FOXA2
Gene Accession No. Q9Y261
Gene Alias FKHL20; forkhead box A2; Forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; RONU; TCF3B; Tcf-3b; Transcription factor 3B; transcription factor Foxa2; WHN
Gene Symbols FOXA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HNF 3 beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3170
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.