Learn More
Invitrogen™ FOXA2 Polyclonal Antibody, Biotin

Description
Additional Information: This antibody is expected to recognise both reported isoforms: NP_068556.2; NP_710141.1. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
Specifications
Specifications
| Antigen | FOXA2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | FOXA2 |
| Gene Accession No. | P32182, Q9Y261 |
| Gene Alias | FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN |
| Gene Symbols | FOXA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.