Learn More
Invitrogen™ FOXA3 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 85%; Rat: 100%; Zebrafish: 79% Immunogen sequence: FENGCYLRRQ KRFKLEEKVK KGGSGAATTT RNGTGSAAST TTPAATVTSP.
Specifications
Specifications
| Antigen | FOXA3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | FOXA3 |
| Gene Accession No. | P35584, P55318 |
| Gene Alias | FKHH3; Fork head-related protein FKH H3; forkhead box A3; forkhead box protein A3; Foxa3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; hepatocyte nuclear factor 3-gamma; HNF3G; HNF-3G; HNF-3-gamma; Tcf3g; Tcf-3g; Transcription factor 3G |
| Gene Symbols | FOXA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.