Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA3 Polyclonal Antibody, Abbomax

Catalog No. 502204099
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4099 Supplier Abbomax Inc Supplier No. 602700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HepG2 cells. Cellular location: Nucleus.

FOXA3 Polyclonal antibody specifically detects FOXA3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FOXA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene FOXA3
Gene Accession No. P32183, P35584, P55318
Gene Alias FKHH3; Fork head-related protein FKH H3; forkhead box A3; forkhead box protein A3; Foxa3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; hepatocyte nuclear factor 3-gamma; HNF3G; HNF-3G; HNF-3-gamma; Tcf3g; Tcf-3g; Transcription factor 3G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of human FOXA3 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15377, 25100, 3171
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.