Learn More
Description
Specifications
Specifications
| Antigen | FOXA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | FKHH3, Fork head-related protein FKH H3, forkhead box A3, Forkhead box protein A3, hepatocyte nuclear factor 3, gamma, hepatocyte nuclear factor 3-gamma, HNF-3G, HNF-3-gamma, HNF3GTCF-3G, MGC10179, TCF3G, Transcription factor 3G |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOXA3 (NP_004488.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
