Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXB1 Antibody (PCRP-FOXB1-1B7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314021AF405
This item is not returnable.
View return policy
Description
FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
FOXB1 | |
Monoclonal | |
Alexa Fluor 405 | |
FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5 | |
Recombinant full-length human FOXB1protein (Uniprot: Q99853) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-FOXB1-1B7 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
27023 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction