Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXB1/FOXB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575259 Supplier Invitrogen™ Supplier No. PA575259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The Forkhead-box (FOX) genes comprise a superfamily of at least 43 members that encode proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOXB1 (forkhead box B1), also known as FKH5 or HFKH-5, and FOXB2 (forkhead box B2) are members of the FOX family and each contain one forkhead DNA-binding domain. Both FOXB1 and FOXB2 localize to the nucleus where they are thought to function as transcription factors that can bind to DNA via their forkhead domains. In mice, defects in the gene encoding FOXB1 are associated with retarded development of the central nervous system (CNS), suggesting that FOXB1 may play a role in CNS organization and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXB1/FOXB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXB2
Gene Accession No. Q5VYV0, Q64732, Q64733, Q99853
Gene Alias bA159H20.4; C43; Fkh4; Fkh5; forkhead box B1; forkhead box B1a; forkhead box B1b; forkhead box B2; forkhead box protein B1; Forkhead box protein B2; forkhead homolog 4; Foxb1; Foxb1a; Foxb1b; FOXB2; Hfh-e5.1; HFKH-5; mesoderm/mesenchyme forkhead 3; Mf3; Transcription factor FKH-4; transcription factor FKH-5; Twh
Gene Symbols FOXB1, FOXB2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 40-87 of Human FoxB1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14240, 27023, 367106, 442425, 64290, 691398
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.