Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590257 Supplier Invitrogen™ Supplier No. PA590257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPNQLTTMGS SLGTGWPHVY GSAGMIDSAT PISMASGDYS AYGVPLKPLC HAAGQTLPAI PVPIKPTPAA VPALPALPAP IPTLLSNSPP SLSPTSSQTA TSQSSPATPS ETLTSPASAL HSVAVH; Positive Samples: HepG2; Cellular Location: Nucleus.

Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Foxb1
Gene Accession No. Q99853
Gene Alias C43; FKH5; forkhead box B1; forkhead box B1a; forkhead box B1b; forkhead box protein B1; Foxb1; Foxb1a; Foxb1b; Hfh-e5.1; HFKH-5; mesoderm/mesenchyme forkhead 3; Mf3; Transcription factor FKH-5; Twh
Gene Symbols Foxb1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human FOXB1 (NP_0363142).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27023
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.