Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXB2 Antibody (PCRP-FOXB2-2B2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314125MFV610
Description
FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow CytometrySpecifications
FOXB2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
442425 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
PCRP-FOXB2-2B2 | |
Flow Cytometry | |
bA159H20.4, forkhead box B2, forkhead box protein B2 | |
Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction