Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564351 Supplier Invitrogen™ Supplier No. PA564351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGRDYKGVLQ AGGLPLASVM HHLGYPVPGQ LGNVVSSVWP HVGVMD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The Forkhead-box (FOX) genes comprise a superfamily of at least 43 members that encode proteins which are involved in transcriptional regulation and maybe associated with the pathogenesis of various cancers. FOXB1 (forkhead box B1), also known as FKH5 or HFKH-5, and FOXB2 (forkhead box B2) are membersof the FOX family and each contain one forkhead DNA-binding domain.Both FOXB1 and FOXB2 localize to the nucleus where they are thought to function as transcription factors that can bind to DNA via their forkhead domains. In mice, defects in the gene encoding FOXB1 are associated with retarded evelopment of the central nervous system (CNS), suggesting that FOXB1 may play a role in CNS organization and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXB2
Gene Accession No. Q5VYV0
Gene Alias bA159H20.4; Fkh4; forkhead box B2; forkhead box protein B2; forkhead homolog 4; Foxb2; Transcription factor FKH-4
Gene Symbols FOXB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXB2. Recombinant protein control fragment (Product #RP-104147).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.