Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse FOXC1, Synthetic Peptide, Invitrogen™

Catalog No. PEP298
Click to view available options
Quantity:
50 μg

Synthetic Peptide

FOXC1 Synthetic Peptide, PEP-298, from Invitrogen™.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it has been shown to play a role in the regulation of embryonic and ocular development. Mutations in this gene cause various glaucoma phenotypes including primary congenital glaucoma, autosomal dominant iridogoniodysgenesis anomaly, and Axenfeld-Rieger anomaly. This peptide corresponds to amino acids A(423) VDDPLPDYSLP (434) of human FOXC1 (corresponds to mouse FOXC1 aa 430-440) and can be used as a blocking peptide with polyclonal antibody PA1-807.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 17300, 2296
Species Human, Mouse
Purification Method purified
Accession Number Q12948, Q61572
Common Name Human/Mouse FOXC1
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Lyophilized
Formulation water with no preservative
For Use With (Application) Blocking Assay, Control
Gene Alias Forkhead box protein C1, FREAC-3
Gene Symbol FOXC1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.