Learn More
Invitrogen™ FOXC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113676
Description
Immunogen sequence: GMGRSYAPYH HQPAAPKDLV KPPYSYIALI TMAIQNAPEK KITLNGIYQF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 100%; Rat: 100%.
Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).
Specifications
| FOXC2 | |
| Polyclonal | |
| Unconjugated | |
| FOXC2 | |
| BF-3; brain factor 3; Fkh14; Fkhl14; forkhead box C2; forkhead box C2 (MFH-1 mesenchyme forkhead 1); forkhead box C2 (MFH-1, mesenchyme forkhead 1); forkhead box protein C2; forkhead homolog 14; forkhead, Drosophila, homolog-like 14; forkhead-related protein FKHL14; FOXC2; Hfhbf3; HFH-BF-3; LD; mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH1; MFH-1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; Transcription factor FKH-14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q61850 | |
| FOXC2 | |
| Synthetic peptide directed towards the N-terminal region of Mouse Foxc2 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.