Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535145
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535145 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535145 Supplier Invitrogen™ Supplier No. PA535145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FOXD1
Gene Accession No. Q16676, Q61345
Gene Alias AI385632; BF-2; brain factor 2; FKHL8; forkhead box D1; forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; forkhead-related protein FKHL8; Forkhead-related transcription factor 4; Foxd1; FREAC4; FREAC-4; Hfh10; Hfhbf2; HFH-BF-2; HNF-3/forkhead homolog, brain factor 2
Gene Symbols FOXD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human FOXD1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15229, 2297
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.