Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA527142

Catalog No. PIPA527142


Description

Description

Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXD1
Polyclonal
Unconjugated
FOXD1
AI385632; BF-2; brain factor 2; FKHL8; forkhead box D1; forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; forkhead-related protein FKHL8; Forkhead-related transcription factor 4; Foxd1; FREAC4; FREAC-4; Hfh10; Hfhbf2; HFH-BF-2; HNF-3/forkhead homolog, brain factor 2
Rabbit
Antigen affinity chromatography
RUO
2297
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q16676
FOXD1
Synthetic peptide corresponding to a region within amino acids 403 and 465 of Human FOXD1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.