Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540424 Supplier Invitrogen™ Supplier No. PA540424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LPARSGPRAP RDVLPHGHEP PAEEAEADLA EDEEESGGCS DGEPRALASR Sequence homology: Cow: 82%; Guinea Pig: 83%; Human: 100%; Mouse: 92%; Rat: 83%.

FOXD2 belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXD2
Gene Accession No. O60548
Gene Alias AI426778; FKHL17; forkhead box D2; forkhead box protein D2; forkhead, drosophila, homolog-like 17; forkhead-like 17; forkhead-related activator 9; forkhead-related protein FKHL17; Forkhead-related transcription factor 9; Foxd2; FREAC9; FREAC-9; Mesoderm/mesenchyme forkhead 2; Mf2; MF-2
Gene Symbols FOXD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FOXD2 (aa 39-88).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2306
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.