Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106981 Supplier Invitrogen™ Supplier No. PA5106981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 86%; Guinea Pig: 86%; Horse: 79%; Human: 79%; Mouse: 100%; Rat: 100%.

FOXD2 belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene FOXD2
Gene Accession No. O35392
Gene Alias AI426778; FKHL17; forkhead box D2; forkhead box protein D2; forkhead, drosophila, homolog-like 17; forkhead-like 17; forkhead-related activator 9; forkhead-related protein FKHL17; Forkhead-related transcription factor 9; Foxd2; FREAC9; FREAC-9; Mesoderm/mesenchyme forkhead 2; Mf2; MF-2
Gene Symbols FOXD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of mouse Foxd2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17301
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.